accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98-10-2

IUPAC Name: benzenesulfonamide
Synonyms: benzenesulfonamide
Molecular Weight (g/mol): 157.19
Molecular Formula: C6H7NO2S
InChi Key: KHBQMWCZKVMBLN-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (4)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (4)
  • (6)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)