accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98-32-8

IUPAC Name: 3-amino-4-hydroxybenzene-1-sulfonamide
Synonyms: 3-amino-4-hydroxybenzenesulfonamide
Molecular Weight (g/mol): 188.2
Molecular Formula: C6H8N2O3S
InChi Key: AVQFHKYAVVQYQO-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1O)S(N)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results