accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98-72-6

IUPAC Name: (4-nitrophenyl)arsonic acid
Synonyms: histostat nitarsone nitarson nitarsonum
Molecular Weight (g/mol): 247.04
Molecular Formula: C6H6AsNO5
InChi Key: FUUFQLXAIUOWML-UHFFFAOYSA-N
SMILES: O[As](O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results