accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH2

CAS RN 98-81-7

IUPAC Name: (1-bromoethenyl)benzene
Synonyms: α-bromostyrene
Molecular Weight (g/mol): 183.05
Molecular Formula: C8H7Br
InChi Key: SRXJYTZCORKVNA-UHFFFAOYSA-N
SMILES: BrC(=C)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)