accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 98-87-3

IUPAC Name: (dichloromethyl)benzene
Synonyms: benzal chloride
Molecular Weight (g/mol): 161.03
Molecular Formula: C7H6Cl2
InChi Key: CAHQGWAXKLQREW-UHFFFAOYSA-N
SMILES: ClC(Cl)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results