accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98-94-2

IUPAC Name: N,N-dimethylcyclohexanamine
Synonyms: dimethylcyclohexylamine
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: SVYKKECYCPFKGB-UHFFFAOYSA-N
SMILES: CN(C)C1CCCCC1
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)