accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 980-71-2

IUPAC Name: (2Z)-but-2-enedioic acid; [3-(4-bromophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
Synonyms: brompheniramine maleate brompheniramine hydrogen maleate
Molecular Weight (g/mol): 435.32
Molecular Formula: C20H23BrN2O4
InChi Key: SRGKFVAASLQVBO-BTJKTKAUNA-N
SMILES: OC(=O)\C=C/C(O)=O.CN(C)CCC(C1=CC=C(Br)C=C1)C1=CC=CC=N1
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)