accessibility menu, dialog, popup

UserName

Viewing profile as user:
INCS

CAS RN 98041-44-2

IUPAC Name: 1-iodo-2-isothiocyanatobenzene
Synonyms: 1-iodo-2-isothiocyanatobenzene
Molecular Weight (g/mol): 261.08
Molecular Formula: C7H4INS
InChi Key: ODOTYLQYSDVPMA-UHFFFAOYSA-N
SMILES: IC1=CC=CC=C1N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results