accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrSSSBr

CAS RN 98057-08-0

IUPAC Name: 5-bromo-5'-(5-bromothiophen-2-yl)-2,2'-bithiophene
Synonyms: 5-bromo-5'-(5-bromothiophen-2-yl)-2,2'-bithiophene
Molecular Weight (g/mol): 406.17
Molecular Formula: C12H6Br2S3
InChi Key: KXFPYYJGYVYXIB-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(Br)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results