CAS RN 98081-83-5
IUPAC Name:
methyl 6-methoxy-1H-indole-2-carboxylate
Synonyms:
methyl 6-methoxy-1H-indole-2-carboxylate
Molecular Weight (g/mol):
205.21
Molecular Formula:
C11H11NO3
InChi Key:
OPUUCOLVBDQWEY-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC2=CC=C(OC)C=C2N1
1
–
1
of
1
results
1
Promotions Available
Methyl 6-methoxyindole-2-carboxylate, 97%
Methyl 6-methoxyindole-2-carboxylate, 97%
CAS: 98081-83-5 Molecular Formula: C11H11NO3 Molecular Weight (g/mol): 205.21 MDL Number: MFCD00134301 InChI Key: OPUUCOLVBDQWEY-UHFFFAOYSA-N Synonym: methyl 6-methoxy-2-indolecarboxylate,6-methoxy-1h-indole-2-carboxylic acid methyl ester,methyl-6-methoxy-2-indolecarboxylate,methyl 6-methoxy-indole-2-carboxylate,6-methoxyindole-2-carboxylic acid methyl ester,methyl 6-methoxyindole-2-carboxylate,1h-indole-2-carboxylic acid, 6-methoxy-, methyl ester,methyl 6-methoxy-2-indolecarboxylat,pubchem7251,6-methoxy-2-indolecarboxylate PubChem CID: 688173 IUPAC Name: methyl 6-methoxy-1H-indole-2-carboxylate SMILES: COC(=O)C1=CC2=CC=C(OC)C=C2N1