accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98167-06-7

IUPAC Name: 2,3-dihydro-1H-indole-2-carboxylic acid
Synonyms: 2,3-dihydro-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: QNRXNRGSOJZINA-UHFFFAOYNA-N
SMILES: OC(=O)C1CC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results