accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHSOOBr

CAS RN 98192-14-4

IUPAC Name: 2-bromo-N-methylbenzene-1-sulfonamide
Synonyms: 2-bromo-N-methylbenzenesulfonamide
Molecular Weight (g/mol): 250.11
Molecular Formula: C7H8BrNO2S
InChi Key: HSCKMNXETNFFAJ-UHFFFAOYSA-N
SMILES: CNS(=O)(=O)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results