accessibility menu, dialog, popup

UserName

Viewing profile as user:
S

CAS RN 98251-31-1

IUPAC Name: 6-phenyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 6-phenyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 260.35
Molecular Formula: C18H12S
InChi Key: BMCNAHBDZUYGJG-UHFFFAOYSA-N
SMILES: S1C2=C(C=CC=C2)C2=C1C(=CC=C2)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results