CAS RN 983-79-9
IUPAC Name:
bis(diphenylmethylidene)hydrazine
Synonyms:
bis(diphenylmethylidene)hydrazine
Molecular Weight (g/mol):
360.46
Molecular Formula:
C26H20N2
InChi Key:
VQSIVLYYQQCXAF-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C(=NN=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Benzophenone azine
CAS: 983-79-9 Molecular Formula: C26H20N2 Molecular Weight (g/mol): 360.46 InChI Key: VQSIVLYYQQCXAF-UHFFFAOYSA-N IUPAC Name: bis(diphenylmethylidene)hydrazine SMILES: C1=CC=C(C=C1)C(=NN=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1