accessibility menu, dialog, popup

UserName

Viewing profile as user:
PP(Z)

CAS RN 983-80-2

IUPAC Name: [(1Z)-2-(diphenylphosphanyl)ethenyl]diphenylphosphane
Synonyms: [(1Z)-2-(diphenylphosphanyl)ethenyl]diphenylphosphane
Molecular Weight (g/mol): 396.41
Molecular Formula: C26H22P2
InChi Key: NCKJIJSEWKIXAT-DQRAZIAOSA-N
SMILES: C(=C\P(C1=CC=CC=C1)C1=CC=CC=C1)\P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)