CAS RN 98349-22-5
CAS RN 98349-22-5
IUPAC Name:
2,4,5-trifluorobenzonitrile
Synonyms:
2,4,5-trifluorobenzonitrile
Molecular Weight (g/mol):
157.1
Molecular Formula:
C7H2F3N
InChi Key:
DLKNOGQOOZFICZ-UHFFFAOYSA-N
SMILES:
FC1=CC(F)=C(C=C1F)C#N
1
–
1
of
1
results
1
2,4,5-Trifluorobenzonitrile 98.0+%, TCI America™
CAS: 98349-22-5 Molecular Formula: C7H2F3N Molecular Weight (g/mol): 157.095 MDL Number: MFCD00013289 InChI Key: DLKNOGQOOZFICZ-UHFFFAOYSA-N Synonym: benzonitrile, 2,4,5-trifluoro,2,4,5-trifluoro-benzonitrile,2,4,5-trifluorobenzenecarbonitrile,3-flurophenol,pubchem1571,acmc-209sa5,2,4,5-trifluorophenylnitrile,ksc488c2t,2,4,5-trifluoro benzonitrile,dlknogqoozficz-uhfffaoysa PubChem CID: 593813 IUPAC Name: 2,4,5-trifluorobenzonitrile SMILES: C1=C(C(=CC(=C1F)F)F)C#N