accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHSBr

CAS RN 98434-22-1

IUPAC Name: 6-bromo-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 6-bromo-2,4-dihydro-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 244.11
Molecular Formula: C8H6BrNOS
InChi Key: QXMFOFHASVVZIT-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(SCC(=O)N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results