accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98475-07-1

IUPAC Name: methyl 2-(bromomethyl)-3-nitrobenzoate
Synonyms: methyl 2-(bromomethyl)-3-nitrobenzoate
Molecular Weight (g/mol): 274.07
Molecular Formula: C9H8BrNO4
InChi Key: FCGIVHSBEKGQMZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(CBr)C(=CC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)