accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98516-76-8

IUPAC Name: (4S)-4-amino-4-[(4-methyl-2-oxo-2H-chromen-7-yl)carbamoyl]butanoic acid
Synonyms: (4S)-4-amino-4-[(4-methyl-2-oxochromen-7-yl)carbamoyl]butanoic acid
Molecular Weight (g/mol): 304.3
Molecular Formula: C15H16N2O5
InChi Key: USALUSONPMPEKY-NSHDSACASA-N
SMILES: CC1=CC(=O)OC2=CC(NC(=O)[C@@H](N)CCC(O)=O)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)