accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98546-51-1

IUPAC Name: [4-(methylsulfanyl)phenyl]boronic acid
Synonyms: 4-(methylsulfanyl)phenylboronic acid
Molecular Weight (g/mol): 168.02
Molecular Formula: C7H9BO2S
InChi Key: IVUHTLFKBDDICS-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)