accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHN(R)OO

CAS RN 98569-12-1

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-1,2,3,4-tetrahydronaphthalene-2-carboxylate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3,4-dihydro-1H-naphthalene-2-carboxylate
Molecular Weight (g/mol): 290.34
Molecular Formula: C16H20NO4
InChi Key: DSDQZWPRZHNCIX-MRXNPFEDSA-M
SMILES: CC(C)(C)OC(=O)N[C@@]1(CCC2=CC=CC=C2C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results