accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)(S)O(S)H3C(R)F(S)OH(S)H3C(R)HOOOH3CO

CAS RN 987-24-6

IUPAC Name: 2-[(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Synonyms: β-val betnovate betamethasone cream flubenisolonvalerate diproderm betamethasone preparation betnelan becort visuβ betamethazone
Molecular Weight (g/mol): 434.5
Molecular Formula: C24H31FO6
InChi Key: AKUJBENLRBOFTD-QZIXMDIESA-N
SMILES: C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(C)=O
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)