CAS RN 98700-50-6
CAS RN 98700-50-6
IUPAC Name:
5-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Synonyms:
5-methyl-1-phenylpyrazole-4-carbaldehyde
Molecular Weight (g/mol):
186.21
Molecular Formula:
C11H10N2O
InChi Key:
IIEFFVFWBUGUTI-UHFFFAOYSA-N
SMILES:
CC1=C(C=O)C=NN1C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 5-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™
CAS: 98700-50-6 Molecular Formula: C11H10N2O Molecular Weight (g/mol): 186.214 InChI Key: IIEFFVFWBUGUTI-UHFFFAOYSA-N PubChem CID: 2776437 IUPAC Name: 5-methyl-1-phenylpyrazole-4-carbaldehyde SMILES: CC1=C(C=NN1C2=CC=CC=C2)C=O