accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98753-19-6

IUPAC Name: 1-({7-[2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl}methyl)-5H,6H,7H-cyclopenta[b]pyridin-1-ium hydrogen sulfate
Synonyms: 1-({7-[2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl}methyl)-5H,6H,7H-cyclopenta[b]pyridin-1-ium hydrogen sulfate
Molecular Weight (g/mol): 612.65
Molecular Formula: C22H24N6O9S3
InChi Key: RKTNPKZEPLCLSF-UHFFFAOYNA-N
SMILES: OS([O-])(=O)=O.CON=C(C(=O)NC1C2SCC(C[N+]3=CC=CC4=C3CCC4)=C(N2C1=O)C(O)=O)C1=CSC(N)=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results