accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNBrNH2

CAS RN 98786-86-8

IUPAC Name: 3-amino-5-bromo-1,2-dihydropyridin-2-one
Synonyms: 3-amino-5-bromo-1H-pyridin-2-one
Molecular Weight (g/mol): 189.01
Molecular Formula: C5H5BrN2O
InChi Key: ULOVLVWNSQNABA-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CNC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results