accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98841-68-0

IUPAC Name: 2-(pyrrolidine-1-carbonyl)phenol
Synonyms: 2-(pyrrolidine-1-carbonyl)phenol
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: LELBEJMWNJWJDG-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results