accessibility menu, dialog, popup

UserName

Viewing profile as user:
NCl

CAS RN 98997-63-8

IUPAC Name: 5λ⁵-azaspiro[4.4]nonan-5-ylium chloride
Synonyms: 5λ⁵-azaspiro[4.4]nonan-5-ylium chloride
Molecular Weight (g/mol): 161.67
Molecular Formula: C8H16ClN
InChi Key: HGAIUEUNFDXNNI-UHFFFAOYSA-M
SMILES: [Cl-].C1CC[N+]2(C1)CCCC2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results