accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-06-9

IUPAC Name: 3-hydroxybenzoic acid
Synonyms: 3-hydroxybenzoic acid acido m-idrossibenzoico kyselina 3-hydroxybenzoova m-salicylic acid m-hba M-hydroxybenzoic acid m-salicylate
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: IJFXRHURBJZNAO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(O)=C1
Percent Purity 
  • (3)
  • (8)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (5)
  • (3)
  • (8)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)