accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHOOHO

CAS RN 99-14-9

IUPAC Name: propane-1,2,3-tricarboxylic acid
Synonyms: tricarballylic acid
Molecular Weight (g/mol): 176.12
Molecular Formula: C6H8O6
InChi Key: KQTIIICEAUMSDG-UHFFFAOYSA-N
SMILES: OC(=O)CC(CC(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)