accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONOONOO

CAS RN 99-34-3

IUPAC Name: 3,5-dinitrobenzoic acid
Synonyms: 3,5-dinitrobenzoic acid
Molecular Weight (g/mol): 212.12
Molecular Formula: C7H4N2O6
InChi Key: VYWYYJYRVSBHJQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)