accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-48-9

IUPAC Name: 2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-ol
Synonyms: carveol laevo carveol p-mentha-6,8-dien-2-ol carveols
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: BAVONGHXFVOKBV-UHFFFAOYNA-N
SMILES: CC(=C)C1CC=C(C)C(O)C1
Boiling Point 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)