accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-62-7

IUPAC Name: 1,3-bis(propan-2-yl)benzene
Synonyms: 1,3-diisopropylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: UNEATYXSUBPPKP-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=CC=C1)C(C)C
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (5)