accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-63-8

IUPAC Name: benzene-1,3-dicarbonyl dichloride
Synonyms: benzene-1,3-dicarbonyl dichloride
Molecular Weight (g/mol): 203.02
Molecular Formula: C8H4Cl2O2
InChi Key: FDQSRULYDNDXQB-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC(=CC=C1)C(Cl)=O
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (6)
  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)