accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-88-7

IUPAC Name: 4-(propan-2-yl)aniline
Synonyms: para-isopropylaniline P-isopropylaniline
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: LRTFPLFDLJYEKT-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(N)C=C1
Boiling Point 
  • (3)
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (1)
  • (5)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)