accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-93-4

IUPAC Name: 1-(4-hydroxyphenyl)ethan-1-one
Synonyms: hydroxyacetophenone piceol usaf KF-15 (4-hydroxyphenyl)ethan-1-one hydroxyacetophenone, para P-hydroxyacetophenone methyl-p-hydroxyphenyl ketone 4-hydroksyacetofenol
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: TXFPEBPIARQUIG-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (6)
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (3)
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (6)
  • (1)