accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99071-54-2

IUPAC Name: 1-(quinolin-6-yl)methanamine
Synonyms: 1-(quinolin-6-yl)methanamine
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: RZIPENSSTUBRAA-UHFFFAOYSA-N
SMILES: NCC1=CC2=CC=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results