accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOOO

CAS RN 99189-60-3

IUPAC Name: 2-[1-(carbamoylmethyl)cyclohexyl]acetate
Synonyms: 2-[1-(carbamoylmethyl)cyclohexyl]acetate
Molecular Weight (g/mol): 198.24
Molecular Formula: C10H16NO3
InChi Key: QJGSJXLCJRXTRY-UHFFFAOYSA-M
SMILES: NC(=O)CC1(CC([O-])=O)CCCCC1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results