accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOOOH

CAS RN 99199-60-7

IUPAC Name: 6-fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid
Synonyms: 6-fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid
Molecular Weight (g/mol): 196.18
Molecular Formula: C10H9FO3
InChi Key: ZNJANLXCXMVFFI-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC2=C(O1)C=CC(F)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results