accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOBrNOO

CAS RN 99277-71-1

IUPAC Name: 4-bromo-2-nitrobenzoate
Synonyms: 4-bromo-2-nitrobenzoate
Molecular Weight (g/mol): 245.01
Molecular Formula: C7H3BrNO4
InChi Key: ZIRHHEZLJGORGU-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(Br)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)