accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHGeH3ClHClHH3C

CAS RN 993-10-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 200.06
Molecular Formula: CH9Cl3Ge
InChi Key: NNMJXXWNUALEKD-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.C[GeH3]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results