accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 993-13-5

IUPAC Name: methylphosphonic acid
Synonyms: methylphosphonic acid
Molecular Weight (g/mol): 96.02
Molecular Formula: CH5O3P
InChi Key: YACKEPLHDIMKIO-UHFFFAOYSA-N
SMILES: CP(O)(O)=O
Quantity 
  • (2)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results