accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOFFFRh

CAS RN 99326-34-8

IUPAC Name: λ¹-rhodium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) trifluoromethanesulfonate
Synonyms: λ¹-rhodium(1+) bis(1,5-cyclooctadiene, (Z,Z)-) triflate
Molecular Weight (g/mol): 468.34
Molecular Formula: C17H24F3O3RhS
InChi Key: GOPBTCMAUMSOBX-QMDOQEJBSA-M
SMILES: [Rh+].[O-]S(=O)(=O)C(F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)