accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHGeHH3CCH3H3C

CAS RN 994-28-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 197.28
Molecular Formula: C6H17ClGe
InChi Key: FJAWDWVPBHGANJ-UHFFFAOYSA-N
SMILES: Cl.CC[GeH](CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results