accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHOOCH3

CAS RN 99469-99-5

IUPAC Name: 2-[3-ethoxy-4-(ethoxycarbonyl)phenyl]acetic acid
Synonyms: [3-ethoxy-4-(ethoxycarbonyl)phenyl]acetic acid
Molecular Weight (g/mol): 252.27
Molecular Formula: C13H16O5
InChi Key: OTGSESBEJUHCES-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(OCC)C=C(CC(O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results