accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiClH3CH3C

CAS RN 995-25-5

IUPAC Name: chlorotripropylsilane
Synonyms: chlorotripropylsilane
Molecular Weight (g/mol): 192.8
Molecular Formula: C9H21ClSi
InChi Key: ACTAPAGNZPZLEF-UHFFFAOYSA-N
SMILES: CCC[Si](Cl)(CCC)CCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)