accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiClH3CH3C

CAS RN 995-45-9

IUPAC Name: tributyl(chloro)silane
Synonyms: tributyl(chloro)silane
Molecular Weight (g/mol): 234.88
Molecular Formula: C12H27ClSi
InChi Key: JSQJUDVTRRCSRU-UHFFFAOYSA-N
SMILES: CCCC[Si](Cl)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)