accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOClN

CAS RN 99586-65-9

IUPAC Name: 4-chloropyridine-2-carboxamide
Synonyms: 4-chloropyridine-2-carboxamide
Molecular Weight (g/mol): 156.57
Molecular Formula: C6H5ClN2O
InChi Key: XIHHOUUTBZSYJH-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(Cl)=CC=N1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)