accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2NH2NOHSOHOO

CAS RN 996-19-0

IUPAC Name: N''-aminoguanidine; sulfuric acid
Synonyms: amino-guanidine; sulfuric acid
Molecular Weight (g/mol): 172.16
Molecular Formula: CH8N4O4S
InChi Key: XLONNWGCEFSFTN-UHFFFAOYSA-N
SMILES: NN=C(N)N.OS(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results