accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NCH3H3C

CAS RN 996-35-0

IUPAC Name: dimethyl(propan-2-yl)amine
Synonyms: N,N-dimethylisopropylamine
Molecular Weight (g/mol): 87.17
Molecular Formula: C5H13N
InChi Key: VMOWKUTXPNPTEN-UHFFFAOYSA-N
SMILES: CC(C)N(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)