accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3SiCH3CH3H3C

CAS RN 996-50-9

IUPAC Name: diethyl(trimethylsilyl)amine
Synonyms: diethyl(trimethylsilyl)amine
Molecular Weight (g/mol): 145.32
Molecular Formula: C7H19NSi
InChi Key: JOOMLFKONHCLCJ-UHFFFAOYSA-N
SMILES: CCN(CC)[Si](C)(C)C
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (4)